Skip to Content
Merck
All Photos(2)

Documents

590444

Sigma-Aldrich

Methyl (4-chlorobenzoyl)acetate

Synonym(s):

(p-Chlorobenzoyl)acetic acid methyl ester, Methyl 3-(4-chlorophenyl)-3-oxopropionate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H9ClO3
CAS Number:
Molecular Weight:
212.63
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

form

solid

mp

42-46 °C (lit.)

SMILES string

COC(=O)CC(=O)c1ccc(Cl)cc1

InChI

1S/C10H9ClO3/c1-14-10(13)6-9(12)7-2-4-8(11)5-3-7/h2-5H,6H2,1H3

InChI key

OIPOJEDRCKVDJK-UHFFFAOYSA-N

Application

Reactant involved in:
  • Oxidative coupling with aminopyridines
  • Enantioselective hydrogenation of unprotected β-enamine esters
  • Intramolecular cyclization
  • Oxidative alkylation of benzylic C-H bonds
  • Tandem oxidative coupling and annulation of phenols

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service