Skip to Content
Merck
All Photos(1)

Key Documents

590274

Sigma-Aldrich

Benzenesulfonyl azide, functionalized silica gel

40-63 μm, extent of labeling: 0.5-1.0 mmol/g loading

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€185.00

€185.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€185.00

About This Item

Linear Formula:
Si(CH2)2C6H4SO2N3
Molecular Weight:
238.32
MDL number:
UNSPSC Code:
12352125
NACRES:
NA.22

€185.00


Check Cart for Availability

Request a Bulk Order

form

solid

Quality Level

reaction suitability

reaction type: solution phase peptide synthesis

extent of labeling

0.5-1.0 mmol/g loading

particle size

40-63 μm

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Since the preparation of the first organic azide, phenyl azide, by Peter Griess in 1864 this energy-rich and versatile class of compounds has enjoyed considerable interest.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service