Skip to Content
Merck
All Photos(1)

Documents

588857

Sigma-Aldrich

3-(Ethoxydimethylsilyl)propylamine

97%

Synonym(s):

(γ-Aminopropyl)dimethylethoxysilane, (3-(Ethoxydimethylsilyl)propyl)amine, (3-Aminopropyl)dimethylethoxysilane, 3-(Ethoxydimethylsilyl)-1-propanamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2OSi(CH3)2(CH2)3NH2
CAS Number:
Molecular Weight:
161.32
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.429 (lit.)

bp

60 °C/8 mmHg (lit.)

density

0.857 g/mL at 25 °C (lit.)

SMILES string

CCO[Si](C)(C)CCCN

InChI

1S/C7H19NOSi/c1-4-9-10(2,3)7-5-6-8/h4-8H2,1-3H3

InChI key

GLISOBUNKGBQCL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

3-(Ethoxydimethylsilyl)propylamine is mainly used as a silane-coupling agent for the surface modification of glass substrates, polymers and nanoparticles via amino silanization.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

136.0 °F - closed cup

Flash Point(C)

57.8 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sonja Petkovic et al.
RNA (New York, N.Y.), 21(7), 1249-1260 (2015-05-23)
Reversible chemistry allowing for assembly and disassembly of molecular entities is important for biological self-organization. Thus, ribozymes that support both cleavage and formation of phosphodiester bonds may have contributed to the emergence of functional diversity and increasing complexity of regulatory
Marguerita Eid et al.
Journal of biophysics (Hindawi Publishing Corporation : Online), 2010, 179641-179641 (2010-01-01)
Ampullosporin A is an antimicrobial, neuroleptic peptaibol, the behavior of which was investigated in different membrane mimetic environments made of egg yolk L-α-phosphatidylcholine. In monolayers, the peptaibol adopted a mixed α/3(10)-helical structure with an in-plane orientation. The binding step was
Rafael C Bernardi et al.
Journal of the American Chemical Society, 141(37), 14752-14763 (2019-08-30)
Can molecular dynamics simulations predict the mechanical behavior of protein complexes? Can simulations decipher the role of protein domains of unknown function in large macromolecular complexes? Here, we employ a wide-sampling computational approach to demonstrate that molecular dynamics simulations, when
Rapid synthesis of dual-responsive hollow capsules with controllable membrane thickness by surface-initiated SET-LRP polymerization.
Liu X, et al.
Macromolecules, 51(3), 1011-1019 (2018)
Lukas F Milles et al.
The journal of physical chemistry. B, 121(15), 3620-3625 (2016-12-20)
The opportunistic pathogen Clostridium perfringens assembles its toxins and carbohydrate-active enzymes by the high-affinity cohesin-dockerin (Coh-Doc) interaction. Coh-Doc interactions characterized previously have shown considerable resilience toward mechanical stress. Here, we aimed to determine the mechanics of this interaction from C.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service