Skip to Content
Merck
All Photos(1)

Key Documents

588369

Sigma-Aldrich

1-Bromopentane-5,5,5-d3

99 atom % D

Synonym(s):

Amyl bromide-5,5,5-d3

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CD3C(CH2)3CH2Br
CAS Number:
Molecular Weight:
154.06
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

isotopic purity

99 atom % D

Assay

99% (CP)

bp

130 °C (lit.)

mp

-95 °C (lit.)

density

1.242 g/mL at 25 °C

mass shift

M+3

SMILES string

[2H]C([2H])([2H])CCCCBr

InChI

1S/C5H11Br/c1-2-3-4-5-6/h2-5H2,1H3/i1D3

InChI key

YZWKKMVJZFACSU-FIBGUPNXSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Flam. Liq. 3 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

89.6 °F - closed cup

Flash Point(C)

32 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service