Skip to Content
Merck
All Photos(1)

Documents

588288

Sigma-Aldrich

1-Bromobutane-4,4,4-d3

98 atom % D

Synonym(s):

Butyl bromide-4,4,4-d3

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CD3(CH2)2CH2Br
CAS Number:
Molecular Weight:
140.04
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:

isotopic purity

98 atom % D

bp

100-104 °C (lit.)

mp

-112 °C (lit.)

density

1.303 g/mL at 25 °C

mass shift

M+3

SMILES string

[2H]C([2H])([2H])CCCBr

InChI

1S/C4H9Br/c1-2-3-4-5/h2-4H2,1H3/i1D3

InChI key

MPPPKRYCTPRNTB-FIBGUPNXSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 2 - Carc. 2 - Flam. Liq. 2 - Repr. 1B - Skin Irrit. 2 - STOT RE 2 Inhalation - STOT SE 3

Target Organs

Liver, Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

71.6 °F - closed cup

Flash Point(C)

22 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yohei Sakaguchi et al.
The Analyst, 140(6), 1965-1973 (2015-02-12)
We have developed a novel amino acid analysis method using derivatization of multiple functional groups (amino, carboxyl, and phenolic hydroxyl groups). The amino, carboxyl, and phenolic hydroxyl groups of the amino acids were derivatized with 1-bromobutane so that the hydrophobicities

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service