Skip to Content
Merck
All Photos(1)

Key Documents

58565

Sigma-Aldrich

Isobutyllithium solution

technical, ~16% in heptane (~1.7 M)

Synonym(s):

Lithium 2-methyl-1-propanide

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
€276.00

€276.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 ML
€276.00

About This Item

Linear Formula:
(CH3)2CHCH2Li
CAS Number:
Molecular Weight:
64.06
Beilstein:
4450775
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

€276.00


Check Cart for Availability

Request a Bulk Order

grade

technical

concentration

~16% in heptane (~1.7 M)

density

0.69 g/mL at 20 °C

storage temp.

2-8°C

SMILES string

[Li]CC(C)C

InChI

1S/C4H9.Li/c1-4(2)3;/h4H,1H2,2-3H3;

InChI key

YZLMQRULZXAMEC-UHFFFAOYSA-N

Signal Word

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1A - STOT SE 3 - Water-react 1

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 2

Flash Point(F)

24.8 °F - closed cup

Flash Point(C)

-4.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service