Skip to Content
Merck
All Photos(1)

Key Documents

579556

Sigma-Aldrich

Ethylene glycol butyl ether

≥99.5%

Synonym(s):

Butyl Cellosolve®, 2-Butoxyethanol, Butyl glycol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3OCH2CH2OH
CAS Number:
Molecular Weight:
118.17
Beilstein:
1732511
EC Number:
MDL number:
UNSPSC Code:
12190000

vapor density

4.1 (vs air)

vapor pressure

<1 mmHg ( 20 °C)
10 mmHg ( 81 °C)

Assay

≥99.5%

autoignition temp.

473 °F

expl. lim.

10.6 %

refractive index

n20/D 1.419 (lit.)

bp

169-172.5 °C (lit.)

mp

−75 °C (lit.)

density

0.902 g/mL at 25 °C (lit.)

SMILES string

CCCCOCCO

InChI

1S/C6H14O2/c1-2-3-5-8-6-4-7/h7H,2-6H2,1H3

InChI key

POAOYUHQDCAZBD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Manufactured by Dow Chemical Company
Cellosolve is a registered trademark of Union Carbide

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

152.6 °F - Pensky-Martens closed cup

Flash Point(C)

67 °C - Pensky-Martens closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Prajakta S Palkar et al.
Toxicology and applied pharmacology, 230(3), 338-345 (2008-04-26)
Administration of a low priming dose of 2-butoxyethanol (BE, 500 mg/kg, p.o.) 7 days prior to a larger LD(90) dose (1500 mg BE/kg, p.o.) offers protection against the lethal dose-induced hemolysis and death in female Sprague Dawley rats because of
G Amir et al.
The British journal of oral & maxillofacial surgery, 49(1), 21-25 (2009-12-26)
Female rats develop haemolytic anaemia and disseminated thrombosis and infarction in multiple organs, including bone, when exposed to 2-butoxyethanol (BE). There is growing evidence that vascular occlusion of the subchondral bone may play a part in some cases of osteoarthritis.
Tawny Hung et al.
Clinical toxicology (Philadelphia, Pa.), 48(6), 569-571 (2010-06-22)
2-butoxyethanol (2BE) is a solvent commonly incorporated into household and industrial cleaning products. Its ingestion causes rapid central nervous system depression, hypotension, and metabolic acidosis attributable to metabolism of the parent compound to butoxyacetic acid (BAA) by alcohol dehydrogenase. Lactic
Annette L Bunge et al.
International journal of pharmaceutics, 435(1), 50-62 (2012-02-15)
Absorption of 2-butoxyethanol (BE) from neat and aqueous solutions of BE was measured through rat skin in vitro and in vivo and through silicone membranes. Like previous studies in human and guinea pig skin, BE flux increased proportional to BE
R Rella et al.
Journal of environmental monitoring : JEM, 14(10), 2659-2662 (2012-08-18)
Indoor air quality is important because people are spending an increasing amount of time in the workplace. They are exposed to outdoor pollutants as well as pollutants emitted from products used indoors. Some chemicals, belonging to the category of volatile

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service