Skip to Content
Merck
All Photos(1)

Key Documents

577847

Sigma-Aldrich

Lead oxide

powder, 99.99% trace metals basis

Synonym(s):

Lead oxide, red, Lead(II,IV) oxide, Minium

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Pb3O4
CAS Number:
Molecular Weight:
685.60
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

vapor pressure

10 mmHg ( 0 °C)

Assay

99.99% trace metals basis

form

powder

SMILES string

O1[PbH2]O[Pb]12O[PbH2]O2

InChI

1S/4O.3Pb

InChI key

XMFOQHDPRMAJNU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Ox. Sol. 2 - Repr. 1A - STOT RE 1

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Fischbein et al.
JAMA, 247(14), 2007-2009 (1982-04-09)
A case of lead poisoning in a female art conservator is reported. The patient had experienced excessive lead exposure while restoring an antique Peruvian tapestry from the Chancay period (1000 to 1500 AD) using a powdered pigment (cinnabar), which had
Yangen Zhou et al.
Inorganic chemistry, 51(23), 12594-12596 (2012-11-13)
Pb(3)O(4) could react with organic dyes in aqueous solution under visible light irradiation, in which Pb(3)O(4) was transformed into Pb(3)(CO(3))(2)(OH)(2) along with oxidation of the organic dyes. Cu(2+) has considerable effect on the reaction. In the presence of Cu(2+), MO
Guo-zhen Lin et al.
Clinica chimica acta; international journal of clinical chemistry, 413(13-14), 1156-1159 (2012-05-09)
Traditional medicines or ethnic remedies, such as Ayurvedic drug of India, have been reported in numerous cases to be one of the main exposure risks for severe lead poisoning. We describe the poisoning of 2 young children from a Chinese
[Acute lead tetroxide poisoning].
K Markiewicz et al.
Polskie Archiwum Medycyny Wewnetrznej, 72(3), 115-121 (1984-09-01)
R D Baer et al.
Journal of ethnopharmacology, 24(1), 31-39 (1988-09-01)
This article discusses the availability of three lead-based salts which are being used as folk remedies in Mexico. Distribution systems and geographic availability were determined, and purchased samples were found to be very high in lead content. The findings suggest

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service