Skip to Content
Merck
All Photos(1)

Key Documents

576743

Sigma-Aldrich

tert-Butyl 3-bromopropionate

97%

Synonym(s):

2-(tert-Butoxycarbonyl)ethyl bromide, 3-Bromopropionic acid tert-butyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H13BrO2
CAS Number:
Molecular Weight:
209.08
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.4470 (lit.)

bp

30 °C/0.2 mmHg (lit.)

density

1.253 g/mL at 25 °C (lit.)

functional group

bromo
ester

storage temp.

2-8°C

SMILES string

CC(C)(C)OC(=O)CCBr

InChI

1S/C7H13BrO2/c1-7(2,3)10-6(9)4-5-8/h4-5H2,1-3H3

InChI key

RMWVUWLBLWBQDS-UHFFFAOYSA-N

Application

tert-Butyl 3-bromopropionate may be used to synthesize the following:
  • tert-butyl-12-cholesteryloxi-4,7,10-trioxadodecanoate[1]
  • 2-(tert-butoxycarbonyl)ethyl 2,3,4-tri-O-acetyl-1-thio-α-L-fucopyranoside[2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

197.6 °F - closed cup

Flash Point(C)

92 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

?Synthesis and characterization of a carboxylic acid derivative for liposomal preparations?
Ionescu C, et al.
Synthesis, 61(04) (2013)
"Synthesis and Biological Evaluation of S-Neofucopeptides as E-and P-Selectin Inhibitors"
Moreno-Vargas.JA, et al.
European Journal of Organic Chemistry, 2008(17), 2973-2982 (2008)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service