Skip to Content
Merck
All Photos(1)

Key Documents

571423

Sigma-Aldrich

1-(2-Methoxyethyl)piperazine

97%

Synonym(s):

4-(2-Methoxyethyl)piperazine, N-(2-Methoxyethyl)piperazine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H16N2O
CAS Number:
Molecular Weight:
144.21
Beilstein:
105098
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.4730 (lit.)

bp

193-194 °C (lit.)

density

0.970 g/mL at 25 °C (lit.)

SMILES string

COCCN1CCNCC1

InChI

1S/C7H16N2O/c1-10-7-6-9-4-2-8-3-5-9/h8H,2-7H2,1H3

InChI key

BMEMBBFDTYHTLH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant for:
  • Preparation of biologically and pharmacologically active molecules

Warning

Air-sensitive - absorbs CO2

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Dam. 1 - Repr. 2 - Skin Corr. 1B - Skin Sens. 1 - STOT RE 1

Target Organs

respiratory tract irritation

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

188.6 °F - closed cup

Flash Point(C)

87 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Son T Nguyen et al.
Bioorganic & medicinal chemistry, 23(9), 2024-2034 (2015-03-31)
Recently we described a novel pyranopyridine inhibitor (MBX2319) of RND-type efflux pumps of the Enterobacteriaceae. MBX2319 (3,3-dimethyl-5-cyano-8-morpholino-6-(phenethylthio)-3,4-dihydro-1H-pyrano[3,4-c]pyridine) is structurally distinct from other known Gram-negative efflux pump inhibitors (EPIs), such as 1-(1-naphthylmethyl)-piperazine (NMP), phenylalanylarginine-β-naphthylamide (PAβN), D13-9001, and the pyridopyrimidine derivatives. Here

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service