Skip to Content
Merck
All Photos(1)

Key Documents

57110

Sigma-Aldrich

Indium(III) chloride hydrate

~39% indium basis, 2-3 mol/mol water

Synonym(s):

Trichloroindium, indium trichloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
InCl3 · aq
CAS Number:
Molecular Weight:
221.18 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.22

reaction suitability

core: indium
reagent type: catalyst

concentration

~39% In

impurities

2-3 mol/mol water

SMILES string

O.Cl[In](Cl)Cl

InChI

1S/3ClH.In.H2O/h3*1H;;1H2/q;;;+3;/p-3

InChI key

KYCHGXYBBUEKJK-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Application

Reagent/catalyst for various organic transformations, e.g. allylation of aldehydes, ketones, and quinones; Diels-Alder reaction; the aldol reaction; Michael additions; the reductive Friedel-Crafts;

Pictograms

Health hazardCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1C - STOT RE 1 Inhalation

Target Organs

Lungs

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Loh, T.-P.; Li, X.-R.,
Angewandte Chemie (International Edition in English), 36, 980-980 (1997)
S Kobayashi et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 6(19), 3491-3494 (2000-11-10)
Group 3-15 metal chlorides (Lewis acids) were classified on the basis of activity and aldehyde- and aldimine-selectivity in an addition reaction of a silyl enol ether. Based on the experimental results, metal chlorides (Lewis acids) were classified as follows: A
Chapman, C. J.; Frost, C. G. et al.
Tetrahedron Letters, 42, 773-773 (2001)
Reddy, B. G.; Kumareswaran, R. et al.
Tetrahedron Letters, 41, 10333-10333 (2000)
Enrique Font-Sanchis et al.
Dalton transactions (Cambridge, England : 2003), (14)(14), 2470-2473 (2009-03-26)
A simple method for the preparation of triorganoindium reagents under mild conditions based in indium-silicon exchange is described.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service