Skip to Content
Merck
All Photos(2)

Key Documents

570214

Sigma-Aldrich

trans-2-Phenylvinylboronic acid pinacol ester

Synonym(s):

(E)-4,4,5,5-Tetramethyl-2-(2-phenylvinyl)-1,3,2-dioxaborolane, (E)-4,4,5,5-Tetramethyl-2-styryl-1,3,2-dioxaborolane, 4,4,5,5-Tetramethyl-2-((E)-2-phenylethenyl)-1,3,2-dioxaborolane, trans-2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)styrene, trans-2-Styreneboronic acid pinacol ester

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€53.30

€53.30


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€53.30

About This Item

Linear Formula:
C6H5CH=CHBO2C2(CH3)4
CAS Number:
Molecular Weight:
230.11
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

€53.30


Check Cart for Availability

Request a Bulk Order

mp

27-33 °C (lit.)

functional group

phenyl

SMILES string

[H]\C(B1OC(C)(C)C(C)(C)O1)=C(\[H])c2ccccc2

InChI

1S/C14H19BO2/c1-13(2)14(3,4)17-15(16-13)11-10-12-8-6-5-7-9-12/h5-11H,1-4H3/b11-10+

InChI key

ARAINKADEARZLZ-ZHACJKMWSA-N

Application

Reactant for:
  • Oxidative coupling reactions catalyzed by areneruthenium
  • Cycloaddition reactions
  • Preparation of allyl vinyl ethers via Cu-catalyzed coupling with alcohols
  • Preparation of 5-vinyl-3-pyridinecarbonitriles as PKCθ inhibitors
  • Diastereoselective addition to zincated hydrazones and stereospecific trapping of boron/zinc bimetallic intermediates by carbon electrophiles
  • Regioselective and stereoselective Pd-catalyzed chelate-controlled intermolecular oxidative Heck reactions

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service