Skip to Content
Merck
All Photos(2)

Key Documents

568708

Sigma-Aldrich

16-Bromohexadecanoic acid

16-Bromohexadecanoic acid

≥99%

Synonym(s):

ψ-Bromohexadecanoic acid, 16-Bromopalmitic acid

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€495.00

€495.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€495.00

About This Item

Linear Formula:
BrCH2(CH2)14CO2H
CAS Number:
Molecular Weight:
335.32
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€495.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥99%

bp

214-217 °C/1 mmHg (lit.)

mp

68-71 °C (lit.)
70-73 °C (lit.)

functional group

bromo
carboxylic acid

SMILES string

OC(=O)CCCCCCCCCCCCCCCBr

InChI

1S/C16H31BrO2/c17-15-13-11-9-7-5-3-1-2-4-6-8-10-12-14-16(18)19/h1-15H2,(H,18,19)

InChI key

PFNCOYVEMJYEED-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

16-Bromohexadecanoic acid can be used:
  • As a starting material in the synthesis of ceramides with ultralong chains.[1]
  • In the synthesis of 6-(nitrooxy)hexadecanoic acid intermediate, which on reacting with (L)-carnitine ethyl ester iodide gave carnitine nitro-derivatives.[2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and characterization of carnitine nitro-derivatives
Piermatti O, et al.
Bioorganic & Medicinal Chemistry, 16(3), 1444-1451 (2008)
Scalable synthesis of human ultralong chain ceramides
Opa?lka L, et al.
Organic Letters, 17(21), 5456-5459 (2015)

Questions

Reviews

Active Filters

  1. 1 Ratings-Only Review

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service