Skip to Content
Merck
All Photos(1)

Key Documents

567701

Sigma-Aldrich

(5-Benzyloxyindol-3-yl)acetonitrile

97%

Synonym(s):

5-Benzyloxyindole-3-acetonitrile, NSC 73391

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H14N2O
CAS Number:
Molecular Weight:
262.31
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

76-80 °C (lit.)

SMILES string

N#CCc1c[nH]c2ccc(OCc3ccccc3)cc12

InChI

1S/C17H14N2O/c18-9-8-14-11-19-17-7-6-15(10-16(14)17)20-12-13-4-2-1-3-5-13/h1-7,10-11,19H,8,12H2

InChI key

ADPRFNVFBYHCJQ-UHFFFAOYSA-N

Application

  • Reactant for preparation of of β-carboline derivatives as antioxidants in the LDL oxidation model
  • Reactant for preparation of indole-N-acetic acid derivatives as aldose reductase inhibitors for diabetic complications treatment
  • Reactant for preparation of tryptamine analogs as antihypertensive agents

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service