Skip to Content
Merck
All Photos(1)

Documents

565032

Sigma-Aldrich

1,4-Dihydroxy-2,6-dimethoxybenzene

97%

Synonym(s):

2,6-Dimethoxyhydroxyquinone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3O)2C6H2(OH)2
CAS Number:
Molecular Weight:
170.16
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

solid

mp

158-162 °C (lit.)

SMILES string

COc1cc(O)cc(OC)c1O

InChI

1S/C8H10O4/c1-11-6-3-5(9)4-7(12-2)8(6)10/h3-4,9-10H,1-2H3

InChI key

GXAVBFNRWXCOPY-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Manickam Selvaraj et al.
Dalton transactions (Cambridge, England : 2003), 48(10), 3291-3299 (2019-02-20)
The major product, 2,3,5-trimethyl-1,4-benzophenone (TMB[double bond, length as m-dash]O), was synthesised by an eco-friendly liquid-phase oxidation of 2,3,6-trimethylphenol (TMP-OH) over Cu(ii)-containing propylsalicylaldimine (CSA) functionalized mesoporous solid catalysts, namely, CSASBA-15(0.2), CSASBA-15(0.1) and CSAMCM-41(0.2), synthesized using various amounts of copper in a
Firoz Shah et al.
Applied and environmental microbiology, 81(24), 8427-8433 (2015-10-04)
Ectomycorrhizal fungi play a key role in mobilizing nutrients embedded in recalcitrant organic matter complexes, thereby increasing nutrient accessibility to the host plant. Recent studies have shown that during the assimilation of nutrients, the ectomycorrhizal fungus Paxillus involutus decomposes organic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service