Skip to Content
Merck
All Photos(1)

Key Documents

56486

Sigma-Aldrich

1-Ethyl-3-methylimidazolium hydrogen sulfate

95%

Synonym(s):

1-Ethyl-3-methylimidazolium hydrosulfate, EMIM HSO4

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
€229.00
1 KG
€1,300.00

€229.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 G
€229.00
1 KG
€1,300.00

About This Item

Empirical Formula (Hill Notation):
C6H12N2O4S
CAS Number:
Molecular Weight:
208.24
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€229.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

95%

SMILES string

OS([O-])(=O)=O.CCn1cc[n+](C)c1

InChI

1S/C6H11N2.H2O4S/c1-3-8-5-4-7(2)6-8;1-5(2,3)4/h4-6H,3H2,1-2H3;(H2,1,2,3,4)/q+1;/p-1

InChI key

HZKDSQCZNUUQIF-UHFFFAOYSA-M

General description

1-Ethyl-3-methylimidazolium hydrogen sulfate is an ionic liquid that is commonly used as a versatile reagent in a variety of applications due to its excellent solvation ability, high thermal stability, and large solubility. It is also used as a solvent and reaction medium in chemical synthesis.

Application

1-Ethyl-3-methylimidazolium hydrogen sulfate [Emim][HSO4] can be used as a reactant to prepare gel polymer electrolyte membranes. By incorporating ionic liquid into the gel polymer matrix, the resulting membranes can exhibit enhanced ionic conductivity and improved electrochemical performance.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nan Shi et al.
Bioresource technology, 283, 174-183 (2019-03-25)
Lignocellulose was oxidatively decomposed in a newly developed polyoxometalates-imidazolium ionic liquid mixture. Aromatic compounds covering acids, esters, ketones, aldehydes, and phenols were selectively produced under various conditions. 4-Hydroxylbenzoic acid was dominatingly yielded under low temperature and high oxidant concentration. Phenolic

Articles

BASIONICS™ Ionic Liquids offer diverse properties for varied applications in the BASF portfolio.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service