Skip to Content
Merck
All Photos(1)

Key Documents

563048

Sigma-Aldrich

2-Bromo-6-methylaniline

95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Br(CH3)C6H3NH2
CAS Number:
Molecular Weight:
186.05
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

refractive index

n20/D 1.6030 (lit.)

bp

105-107 °C/205 mmHg (lit.)

density

1.4780 g/mL at 25 °C (lit.)

functional group

bromo

SMILES string

Cc1cccc(Br)c1N

InChI

1S/C7H8BrN/c1-5-3-2-4-6(8)7(5)9/h2-4H,9H2,1H3

InChI key

LDUCMSVRKKDATH-UHFFFAOYSA-N

Application

2-Bromo-6-methylaniline can be used as a reactant to synthesize:
  • Alkyl substituted bromoindazole building blocks by Pd-catalyzed Suzuki coupling reaction with various vinyl boronic acids.[1]
  • Difluoropyridoindole via Pd-catalyzed intramolecular Heck reaction with difluoropiperidinone.[2]
  • 4-Methyl-N-phenyl-1H-benzo[d]imidazol-2-amine by one-pot Cu-catalyzed domino C-N cross-coupling reaction with iodobenzene and thiourea.[3]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

215.1 °F - closed cup

Flash Point(C)

101.7 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Copper-promoted one-pot approach: Synthesis of benzimidazoles
Boddapati SN, et al.
Molecules (Basel), 25(8), 1788-1788 (2020)
Novel synthesis of 4, 4-difluoropyrido [4, 3-b] indoles via intramolecular Heck reaction
Madaiah M, et al.
Tetrahedron Letters, 54(11), 1424-1427 (2013)
Synthesis and evaluation of indazole based analog sensitive Akt inhibitors
Okuzumi T, et al.
Molecular Biosystems, 6(8), 1389-1402 (2010)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service