Skip to Content
Merck
All Photos(1)

Key Documents

562416

Sigma-Aldrich

Bis(tetramethylcyclopentadienyl)iron(II)

Synonym(s):

Octamethylferrocene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H26Fe
CAS Number:
Molecular Weight:
298.24
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

form

solid

reaction suitability

core: iron
reagent type: catalyst

mp

120-160 °C (lit.)

storage temp.

2-8°C

SMILES string

[Fe].C[C]1[CH][C](C)[C](C)[C]1C.C[C]2[CH][C](C)[C](C)[C]2C

InChI

1S/2C9H13.Fe/c2*1-6-5-7(2)9(4)8(6)3;/h2*5H,1-4H3;

InChI key

SDTGEKAAGAIGJL-UHFFFAOYSA-N

Application

  • Catalyst for alkyd paint drying via mediation of auto-oxidation reactions
  • Electron donor in photoinduced electron-transfer reduction reactions controlled by conformational distortion
  • Electrocatalytical hydrogen peroxide and glucose sensor
  • Used for inter- and intramolecular quenching of the lowest singlet excited state of porphyrins

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service