Skip to Content
Merck
All Photos(2)

Key Documents

560278

Sigma-Aldrich

5-Aminoisatoic anhydride

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€155.00

€155.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€155.00

About This Item

Empirical Formula (Hill Notation):
C8H6N2O3
CAS Number:
Molecular Weight:
178.14
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€155.00


Check Cart for Availability

Request a Bulk Order

form

solid

Quality Level

impurities

may contain ≤10% DMF

mp

>300 °C (lit.)

functional group

ester

SMILES string

Nc1ccc2NC(=O)OC(=O)c2c1

InChI

1S/C8H6N2O3/c9-4-1-2-6-5(3-4)7(11)13-8(12)10-6/h1-3H,9H2,(H,10,12)

InChI key

QEQDLBKVHJXPJA-UHFFFAOYSA-N

Application

Reactant for:
  • β-cyclodextrin catalyzed synthesis of tryptanthrin and its analogs in water
  • Synthesis of amino-chloroquinolyl derivatives as antimalarial agents

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and antimalarial activity of new 4-amino-7-chloroquinolyl amides, sulfonamides, ureas and thioureas.
Ekoue-Kovi K, et al.
Bioorganic & Medicinal Chemistry, 17(1), 270-283 (2009)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service