Skip to Content
Merck
All Photos(1)

Key Documents

560073

Sigma-Aldrich

9,9-Dioctyl-2,7-dibromofluorene

96%

Synonym(s):

2,7-Dibromo-9,9-bis(n -octyl)fluorene, 2,7-Dibromo-9,9-di-n -octyl-9H -fluorene, 2,7-Dibromo-9,9-di-n -octylfluorene, 2,7-Dibromo-9,9-dioctyl-9H -fluorene, 9,9-Dioctyl-2,7-dibromofluorene, 9,9-Dioctyl-9H -2,7-dibromofluorene

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€257.00

€257.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
€257.00

About This Item

Empirical Formula (Hill Notation):
C29H40Br2
CAS Number:
Molecular Weight:
548.44
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

€257.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

96%

form

solid

mp

59-63 °C (lit.)

SMILES string

CCCCCCCCC1(CCCCCCCC)c2cc(Br)ccc2-c3ccc(Br)cc13

InChI

1S/C29H40Br2/c1-3-5-7-9-11-13-19-29(20-14-12-10-8-6-4-2)27-21-23(30)15-17-25(27)26-18-16-24(31)22-28(26)29/h15-18,21-22H,3-14,19-20H2,1-2H3

InChI key

CYKLQIOPIMZZBZ-UHFFFAOYSA-N

General description

Intermediate for polymeric light-emitting diodes.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shotaro Hayashi et al.
Scientific reports, 7(1), 1078-1078 (2017-04-26)
Absolute molecular weight values obtained by gel permeation chromatography with multi angle light scattering of PEDOTF8 were approximately 65% of the relative molecular weight values obtained by gel permeation chromatography using polystyrene standards. Both light absorption and molecular weight measurements
Zhiwei Chu et al.
Sensors (Basel, Switzerland), 18(5) (2018-05-16)
An aggregation-caused quenching (ACQ)-active polymer (PF), an aggregation-induced emission (AIE)-active polymer (PFTPE) and an aggregation-enhanced emission (AEE)-active polymer (PTTPE) were synthesized by tetraphenylethane (TPE), fluorene and thiophene moieties. Polyurethane (PU) foams modified by PF, PFTPE and PTTPE, namely PU-PF, PU-PFTPE

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service