Skip to Content
Merck
All Photos(1)

Documents

55940

Sigma-Aldrich

3-Hydroxy-2-naphthoic hydrazide

≥98.0%

Synonym(s):

3-Hydroxy-2-naphthydrazide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC10H6CONHNH2
CAS Number:
Molecular Weight:
202.21
Beilstein:
395153
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥98.0%

mp

205-208 °C (lit.)
205-208 °C

SMILES string

NNC(=O)c1cc2ccccc2cc1O

InChI

1S/C11H10N2O2/c12-13-11(15)9-5-7-3-1-2-4-8(7)6-10(9)14/h1-6,14H,12H2,(H,13,15)

InChI key

FDNAQCWUERCJBK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

3-Hydroxy-2-naphthoic hydrazide may be used to synthesize the following 2-(alkyl/arylamino)-5-(3-hydroxy-2-naphthyl)-1,3,4-thiadiazoles:
  • 2-ethylamino-5-(3-hydroxy-2-naphthyl)-1,3,4-thiadiazole
  • 2-phenethylamino-5-(3-hydroxy-2-naphthyl)-1,3,4-thiadiazole
  • 2-phenylamino-5-(3-hydroxy-2-naphthyl)-1,3,4-thiadiazole
  • 2-(p-bromophenylamino)-5-(3-hydroxy-2-naphthyl)-1,3,4-thiadiazole
  • 2-(p-chlorophenylamino)-5-(3-hydroxy-2-naphthyl)-1,3,4-thiadiazole
  • 2-(p-fluorophenylamino)-5-(3-hydroxy-2-naphthyl)-1,3,4-thiadiazole
  • 2-(m-fluorophenylamino)-5-(3-hydroxy-2-naphthyl)-1,3,4-thiadiazole
  • 2-(p-methoxyphenylamino)-5-(3-hydroxy-2-naphthyl)-1,3,4-thiadiazole
  • 2-(p-methylphenylamino)-5-(3-hydroxy-2-naphthyl)-1,3,4-thiadiazole
  • 2-(m-trifluoromethylphenylamino)-5-(3-hydroxy-2-naphthyl)-1,3,4-thiadiazole
It may also be employed for the synthesis of 3-hydroxy-2-naphthaldehyde.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Histochemical demonstration of protein-bound amino groups.
L P WEISS et al.
The journal of histochemistry and cytochemistry : official journal of the Histochemistry Society, 2(1), 29-49 (1954-01-01)
A Pompella et al.
The American journal of pathology, 142(5), 1353-1357 (1993-05-01)
Confocal laser scanning fluorescence microscopy plus image videoanalysis was used to visualize the tissue areas and the subcellular sites first involved by oxidative stress and lipid peroxidation, in the well-established experimental model of lipid peroxidation induced by haloalkane intoxication in
Molecular structure, vibrational spectroscopic, first-order hyperpolarizability and HOMO, LUMO studies of 3-hydroxy-2-naphthoic acid hydrazide.
Karpagam J, et al.
Journal of Raman Spectroscopy, 41(1), 53-62 (2010)
Hatice N Dogan et al.
Bioorganic & medicinal chemistry, 10(9), 2893-2898 (2002-07-12)
Two new series of 2,5-disubstituted-1,3,4-thiadiazoles were synthesized for their possible anticonvulsant, antibacterial and antifungal activities. The degree of protection afforded by these compounds at a dose of 100mg/kg i.p. against pentylenetetrazole-induced convulsions in mice ranged from 0 to 90%. Among
G Nöhammer
Histochemistry, 94(5), 485-488 (1990-01-01)
Fixed cells and tissues pretreated with 4-hydroxynonenal were used as models for the histochemical demonstration of protein bound aldehydic groups. The aldehydes were stained with both a modification of the 2,4-dinitrophenylhydrazine method (2,4-DNPH) and the optimized staining using 3-hydroxy-2-naphthoic acid

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service