Skip to Content
Merck
All Photos(1)

Documents

559245

Sigma-Aldrich

Ethyl (3-trifluoromethylbenzoyl)acetate

97%

Synonym(s):

3-(3-Trifluoromethylphenyl)-3-oxo-propionic acid ethyl ester, Ethyl 2-(3-trifluoromethylbenzoyl)acetate, Ethyl 3-(3-trifluoromethylphenyl)-3-oxopropanoate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3C6H4COCH2CO2C2H5
CAS Number:
Molecular Weight:
260.21
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.4850 (lit.)

bp

250-251 °C (lit.)

density

1.267 g/mL at 25 °C (lit.)

SMILES string

CCOC(=O)CC(=O)c1cccc(c1)C(F)(F)F

InChI

1S/C12H11F3O3/c1-2-18-11(17)7-10(16)8-4-3-5-9(6-8)12(13,14)15/h3-6H,2,7H2,1H3

InChI key

YCHPVUWFIZXXPI-UHFFFAOYSA-N

Application

Reactant for:
  • Preparation of pyrido[1,2-a]benzimidazoles as cytotoxic and antiplasmodial agent
  • Preparation of diaryl-substituted pyrazoles as potent CCR2 receptor antagonists
  • Acid-catalyzed addition/annulation reactions
  • Cyclocondensation with diaminophenylpyrazole

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service