Skip to Content
Merck
All Photos(1)

Key Documents

558850

Sigma-Aldrich

[(S)-(−)-1-(4-Nitrophenyl)-2-pyrrolidinemethyl]acrylate

97%

Synonym(s):

NPP acrylate

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€280.00

€280.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€280.00

About This Item

Empirical Formula (Hill Notation):
C14H16N2O4
CAS Number:
Molecular Weight:
276.29
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

€280.00


Check Cart for Availability

Request a Bulk Order

Assay

97%

optical activity

[α]20/D −46.0°, c = 1% in methanol

contains

500 ppm TBC as inhibitor

mp

38-48 °C (lit.)

storage temp.

2-8°C

SMILES string

[O-][N+](=O)c1ccc(cc1)N2CCC[C@H]2COC(=O)C=C

InChI

1S/C14H16N2O4/c1-2-14(17)20-10-13-4-3-9-15(13)11-5-7-12(8-6-11)16(18)19/h2,5-8,13H,1,3-4,9-10H2/t13-/m0/s1

InChI key

HCVPUFHAWIINEQ-ZDUSSCGKSA-N

General description

NLO monomer

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service