Skip to Content
Merck
All Photos(1)

Key Documents

558052

Sigma-Aldrich

(R)-(−)-4-Methyl-2-pentanol

97%, optical purity99% (ee) (GLC)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCH2CH(OH)CH3
CAS Number:
Molecular Weight:
102.17
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:

Assay

97%

optical activity

[α]22/D −18.5°, c = 1% in chloroform

optical purity

99% (ee) (GLC)

refractive index

n20/D 1.4630 (lit.)

bp

132 °C (lit.)

density

0.802 g/mL at 25 °C (lit.)

SMILES string

CC(C)C[C@@H](C)O

InChI

1S/C6H14O/c1-5(2)4-6(3)7/h5-7H,4H2,1-3H3/t6-/m1/s1

InChI key

WVYWICLMDOOCFB-ZCFIWIBFSA-N

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Flam. Liq. 3 - STOT SE 3

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

105.8 °F - closed cup

Flash Point(C)

41 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H Tomczyk et al.
Medycyna pracy, 30(6), 417-423 (1979-01-01)
A gas chromatographic method of determination of methyl isobutyl ketone, methyl isobutyl carbinol, acetone, toluene and o-xylene has been elaborated. Those compounds have been best separated on the column containing 10% Carbowax 20 M on Chromosorb W. Air samples were
Chi M Phan et al.
The journal of physical chemistry. B, 116(3), 980-986 (2011-12-17)
The surface potential (ΔV) and surface tension (γ) of MIBC (methyl isobutyl carbinol) were measured on the subphase of pure water and electrolyte solutions (NaCl at 0.02 and 2 M). In contrast to ionic surfactants, it was found that surface
M Vézina et al.
Canadian journal of physiology and pharmacology, 68(8), 1055-1061 (1990-08-01)
The hepatonecrogenic properties of chloroform (CHCl3) can be modified by the administration of various chemicals. The ability of methyl isobutyl ketone (MIBK) and its two major metabolites, 4-methyl-2-pentanol (4MPOL) and 4-hydroxymethyl isobutyl ketone (4-OHMIBK) to potentiate the liver injury induced
Andrés M Castillo et al.
Journal of magnetic resonance (San Diego, Calif. : 1997), 209(2), 123-130 (2011-02-15)
The computational cost for the simulation of NMR spectra grows exponentially with the number of nuclei. Today, the memory available to store the Hamiltonian limits the size of the system that can be studied. Modern computers enable to tackle systems
Ralph Gingell et al.
Toxicology letters, 136(3), 199-204 (2002-12-31)
Methyl isobutyl carbinol (MIBC) is an oxygenated solvent that is metabolized to methylisobutyl ketone (MIBK) and then to 4-hydroxymethyl-4-methyl-2-pentanone (HMP). Plasma levels of MIBC, MIBK and HMP were determined up to 12 h after a single oral 5 mmol/kg dose

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service