Skip to Content
Merck
All Photos(2)

Key Documents

556386

Sigma-Aldrich

Polyaniline (emeraldine base)

average Mw ~50,000

Synonym(s):

Emeraldine base polyaniline

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€169.00
25 G
€822.00

€169.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€169.00
25 G
€822.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

€169.00


Check Cart for Availability

Request a Bulk Order

mol wt

average Mw ~50,000

Quality Level

solubility

m-cresol: soluble
DMAC: soluble
DMF: soluble
DMSO: soluble
NMP: soluble

λmax

~330 nm

SMILES string

Brc1nn[nH]c1C.Cl

InChI

1S/C3H4BrN3.ClH/c1-2-3(4)6-7-5-2;/h1H3,(H,5,6,7);1H

InChI key

LYCWKZFKTMHXQA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Polyaniline (PAni) emeraldine salt can be prepared by protonation of PAni emeraldine base (PAni-EB). PAni-EB forms at pH>7. Its mechanism against corrosive agents has been explained in a study.[1]

Application

PAni-EB may be used as a substrate for graphene transfer.[2]

Features and Benefits

Undoped form of conducting polymer (non-conductive).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Annealing free, clean graphene transfer using alternative polymer scaffolds.
Wood JD, et al.
Nanotechnology, 26(5) (2015)
Study of epoxy and alkyd coatings modified with emeraldine base form of polyaniline.
Armelin E, et al.
Progress in Organic Coatings, 58, 316-322 (2007)
Jia Liu et al.
Science (New York, N.Y.), 367(6484), 1372-1376 (2020-03-21)
The structural and functional complexity of multicellular biological systems, such as the brain, are beyond the reach of human design or assembly capabilities. Cells in living organisms may be recruited to construct synthetic materials or structures if treated as anatomically

Articles

Self-healing soft electronic materials offer potential cost savings and reduced electronic waste.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service