556149
4-Isopropylbenzenesulfonyl chloride
technical grade
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
grade
technical grade
form
solid
refractive index
n20/D 1.5410 (lit.)
density
1.220 g/mL at 25 °C (lit.)
SMILES string
CC(C)c1ccc(cc1)S(Cl)(=O)=O
InChI
1S/C9H11ClO2S/c1-7(2)8-3-5-9(6-4-8)13(10,11)12/h3-7H,1-2H3
InChI key
CETRNHJIXGITKR-UHFFFAOYSA-N
General description
4-Isopropylbenzenesulfonyl chloride can be synthesized from isopropylbenzene via chlorosulfonation.
Application
4-Isopropylbenzenesulfonyl chloride may be used to synthesize chlorosulfonyl-functionalized ATRP (Atom transfer radical polymerization) initiator and 4-isopropylbenzenethiol.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Skin Corr. 1B
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Original route to polylactide-polystyrene diblock copolymers containing a sulfonyl group at the junction between both blocks as precursors to functional nanoporous materials.
Reactive functional Polymers, 72(8), 495-502 (2012)
The Journal of organic chemistry, 71(16), 6248-6250 (2006-07-29)
The ring inversion barrier for thianthrene tetraoxide was determined by making use of the variable temperature 13C NMR spectra of the 2,7-diisopropyl derivative (DeltaG(double dagger) = 6.5 kcal mol(-1)). The barrier is lower than that measured for a trans thianthrene
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service