Skip to Content
Merck
All Photos(1)

Key Documents

553891

Sigma-Aldrich

1-Bromo-2-(trifluoromethoxy)benzene

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrC6H4OCF3
CAS Number:
Molecular Weight:
241.01
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.464 (lit.)

bp

158-160 °C (lit.)

density

1.62 g/mL at 25 °C (lit.)

SMILES string

FC(F)(F)Oc1ccccc1Br

InChI

1S/C7H4BrF3O/c8-5-3-1-2-4-6(5)12-7(9,10)11/h1-4H

InChI key

ITYCJCVRPBLODP-UHFFFAOYSA-N

General description

1-Bromo-2-(trifluoromethoxy)benzene can be prepared from (trifluoromethoxy)benzene.

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

140.0 °F - closed cup

Flash Point(C)

60 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

2-, 3-, and 4-(Trifluoromethoxy) phenyllithiums: Versatile Intermediates Offering Access to a Variety of New Organofluorine Compounds.
Castagnetti E and Schlosser M.
European Journal of Organic Chemistry, 4, 691-695 (2001)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service