Skip to Content
Merck
All Photos(2)

Key Documents

553778

Sigma-Aldrich

9-Ethyl-3-nitrocarbazole

98%

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€394.00

€394.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
€394.00

About This Item

Empirical Formula (Hill Notation):
C14H12N2O2
CAS Number:
Molecular Weight:
240.26
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€394.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

98%

mp

128-130 °C (lit.)

functional group

nitro

SMILES string

CCn1c2ccccc2c3cc(ccc13)[N+]([O-])=O

InChI

1S/C14H12N2O2/c1-2-15-13-6-4-3-5-11(13)12-9-10(16(17)18)7-8-14(12)15/h3-9H,2H2,1H3

InChI key

WONHLSYSHMRRGO-UHFFFAOYSA-N

General description

9-Ethyl-3-nitrocarbazole can be prepared from 9-ethyl carbazole via nitration.[1] Its crystals exhibit triclinic crystal system.[2]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Structures of 9-ethyl-3-nitrocarbazole and 1-(9-carbazolyl)-3-(3-nitro-9-carbazolyl) propane.
Chen Z, et al.
Acta Crystallographica Section C, Structural Chemistry, 48(12), 2185-2189 (1992)
Chlorination of carbazole and its derivatives with 1-chlorobenzotriazole.
Bowyer PM, et al.
J. Chem. Soc. Sect. C, 2775-2777 (1971)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service