Skip to Content
Merck
All Photos(1)

Key Documents

553077

Sigma-Aldrich

4-Pentenenitrile

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2=CHCH2CH2CN
CAS Number:
Molecular Weight:
81.12
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Assay:
97%

Assay

97%

refractive index

n20/D 1.42 (lit.)

bp

140 °C (lit.)

density

0.814 g/mL at 25 °C (lit.)

SMILES string

C=CCCC#N

InChI

1S/C5H7N/c1-2-3-4-5-6/h2H,1,3-4H2

InChI key

CFEYBLWMNFZOPB-UHFFFAOYSA-N

General description

4-Pentenenitrile (4-PN) is a terminal alkene nitrile. It is obtained from 3-pentenenitrile via cationic nickel hydride or cobalt catalyzed isomerization. 4-Pentenenitrile undergoes hydrocyanation in the presence of bidentate nickel complexes (catalysts) to yield 3-pentenenitrile. 4-PN undergoes gas-phase reaction with OH radicals and Cl atoms in the presence of synthetic air and various reference compounds.

Application

4-Pentenenitrile may be used to synthesize 4-pentenylamine.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

B Ozierenski et al.
Die Nahrung, 37(1), 5-14 (1993-01-01)
The effects of 5-vinyloxazolidine-2-thione (VOT), 1-cyano-3-butene (CYB) and various isothiocyanates on parameters of hepatic phase I and phase II biotransformation were investigated in male rats after oral treatment for 3 consecutive days. The compounds with the exception of CYB caused
Ligand Descriptor Analysis in Nickel-Catalysed Hydrocyanation: A Combined Experimental and Theoretical Study.
Burello E, et al.
Advanced Synthesis & Catalysis, 347(6), 803-810 (2005)
Gas-phase reactivity study of (E)-3-pentenenitrile and 4-pentenenitrile towards OH radicals and Cl atoms at atmospheric pressure.
Colomer JP, et al.
Atmospheric Environment, 61, 597-604 (2012)
New methods for alkaloid synthesis: Generation of indole-2, 3-diquinomethanes as a route to indole alkaloids.
Gallagher T and Magnus P.
Tetrahedron, 37(23), 3889-3897 (1981)
P Kloss et al.
Poultry science, 73(10), 1542-1551 (1994-10-01)
Glucosinolates and their breakdown products (nitriles) have long been implicated as toxic factors when feeding rapeseed (Brassica napus) meals and crambe (Crambe abyssinica) meals to poultry. Accordingly, various methods have been developed to remove these compounds from the meals to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service