Skip to Content
Merck
All Photos(1)

Documents

550604

Sigma-Aldrich

4-Chloromethyl-2,2-dimethyl-1,3-dioxolane

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H11ClO2
CAS Number:
Molecular Weight:
150.60
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.434 (lit.)

bp

156-158 °C (lit.)

density

1.063 g/mL at 25 °C (lit.)

SMILES string

CC1(C)OCC(CCl)O1

InChI

1S/C6H11ClO2/c1-6(2)8-4-5(3-7)9-6/h5H,3-4H2,1-2H3

InChI key

BNPOTXLWPZOESZ-UHFFFAOYSA-N

Related Categories

General description

4-Chloromethyl-2,2-dimethyl-1,3-dioxolane is a ketal that can be synthesized by the reaction of 3-chloro-propane-1,2-diol (3-MCPD) with acetone in the presence of p-toluene sulfonic acid.

Application

4-Chloromethyl-2,2-dimethyl-1,3-dioxolane may be used in the preparation of 2,2-dimethyl-4-methylene-1,3-dioxolane via dehydrochlorination.

Pictograms

FlameSkull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

127.9 °F - closed cup

Flash Point(C)

53.3 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

On the Cationic Polymerization of 2, 2-Dimethyl-1, 3-dioxole and 2,2-Diphenyl-1,3-dioxole.
Akkapeddi MK and Reimschuessel HK.
Macromolecules, 2(5), 827-831 (1979)
Occurrence of 3-chloro-propane-1, 2-diol (3-MCPD) and related compounds in foods: a review.
Hamlet CG, et al.
Food Additives and Contaminants, 19(7), 619-631 (2002)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service