Skip to Content
Merck
All Photos(1)

Documents

550477

Sigma-Aldrich

3,9-Dodecadiyne

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2C≡C(CH2)4C≡CCH2CH3
CAS Number:
Molecular Weight:
162.27
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

refractive index

n20/D 1.465 (lit.)

bp

55-58 °C/0.5 mmHg (lit.)

density

0.809 g/mL at 25 °C (lit.)

SMILES string

CCC#CCCCCC#CCC

InChI

1S/C12H18/c1-3-5-7-9-11-12-10-8-6-4-2/h3-4,9-12H2,1-2H3

InChI key

SLQXYOMUNKJHRV-UHFFFAOYSA-N

General description

3,9-Dodecadiyne is an internal diyne that can undergo nickel-catalyzed cycloaddition with aryl nitriles to form pyridines.

Application

3,9-Dodecadiyne may be used as a monomer to prepare soluble hyperbranched poly(phenylenealkenes) via transition metal-catalyzed polycyclotrimerization. It may also be used as a starting material for preparing 1,2,3,4,9,10-hexaethyl-6,7,8,9-tetrahydroanthracene.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

New catalysts for polymerizations of substituted acetylenes.
Tang BZ, et al.
ACS Symp. Ser., 760 (2000)
A nickel-catalyzed route to pyridines.
McCormick MM, et al.
Journal of the American Chemical Society, 127(14), 5030-5031 (2005)
Novel synthetic route to octasubstituted naphthalenes from four alkynes and one olefin unit via zirconacyclopentadienes and 1,2-diiodo-3,4,5,6-tetraalkylbenzene.
Zhou X, et al.
The Journal of Organic Chemistry, 69(13), 4559-4562 (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service