Skip to Content
Merck
All Photos(1)

Key Documents

550450

Sigma-Aldrich

4-Bromo-4′-fluorobiphenyl

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrC6H4C6H4F
CAS Number:
Molecular Weight:
251.09
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

92.5-97.5 °C (lit.)

functional group

bromo
fluoro

SMILES string

Fc1ccc(cc1)-c2ccc(Br)cc2

InChI

1S/C12H8BrF/c13-11-5-1-9(2-6-11)10-3-7-12(14)8-4-10/h1-8H

InChI key

XFGPSHWWPIFPNL-UHFFFAOYSA-N

General description

4-Bromo-4′-fluorobiphenyl is a 4,4′-disubstituted biphenyl derivative that can be prepared by reacting 4-fluorobenzeneboronic acid with 4-bromo-iodobenzene. Its crystals belong to the monoclinic crystal system and P21/c space group.

Application

4-Bromo-4′-fluorobiphenyl may be used in the preparation of 5,5′-bis(4′-fluoro-biphenyl-4-yl)-2,2′:5′,2′-terthiophene (p-F2BP3T), a fluorinated inducing layer for weak epitaxy growth (WEG).

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Weak epitaxy growth of phthalocyanine on inducing layers of fluorinated 5,5?-Bis(biphenyl-4-yl)-2,2':5',2?-terthiophene.
Qiao X, et al.
The Journal of Physical Chemistry B, 116(6), 1812-1818 (2012)
Structure of 4-bromo-4'-fluorobiphenyl.
Gleason WB, et al.
Acta Crystallographica Section C, Structural Chemistry, 47(7), 1473-1476 (1991)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service