Skip to Content
Merck
All Photos(1)

Key Documents

549746

Sigma-Aldrich

Tetrahydro-2-methyl-3-thioxo-1,2,4-triazine-5,6-dione

97%

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€394.00

€394.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
€394.00

About This Item

Empirical Formula (Hill Notation):
C4H5N3O2S
CAS Number:
Molecular Weight:
159.17
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€394.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

mp

168-171 °C (lit.)

functional group

thiourea

SMILES string

CN1NC(=O)C(=O)NC1=S

InChI

1S/C4H5N3O2S/c1-7-4(10)5-2(8)3(9)6-7/h1H3,(H,6,9)(H,5,8,10)

InChI key

UMWWHOXOVPIGFD-UHFFFAOYSA-N

General description

Tetrahydro-2-methyl-3-thioxo-1,2,4-triazine-5,6-dione is an intermediate for synthesizing ceftriaxone.[1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mio Tange et al.
Chemical & pharmaceutical bulletin, 64(3), 207-214 (2016-03-05)
Ceftriaxone sodium preparation for injection is known to form insoluble microparticles with calcium. The purpose of this study was to evaluate the role of an impurity in the ceftriaxone sodium preparation on this incompatibility. Firstly, using HPLC, two impurities were

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service