Skip to Content
Merck
All Photos(1)

Key Documents

548065

Sigma-Aldrich

2-Methyl-1-naphthaldehyde

97%

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€289.00

€289.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€289.00

About This Item

Linear Formula:
CH3C10H6CHO
CAS Number:
Molecular Weight:
170.21
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€289.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

form

solid

mp

51-56 °C (lit.)

functional group

aldehyde

storage temp.

2-8°C

SMILES string

Cc1ccc2ccccc2c1C=O

InChI

1S/C12H10O/c1-9-6-7-10-4-2-3-5-11(10)12(9)8-13/h2-8H,1H3

InChI key

WIAZTPUQHUFMQA-UHFFFAOYSA-N

General description

2-Methyl-1-naphthaldehyde can be prepared from 1-(bromomethyl)-2-methylnaphthalene.[1]

Application

2-Methyl-1-naphthaldehyde may be used in the preparation of 2-methyl-1-naphthaldehyde tosylhydrazone.[2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Carbene-carbene rearrangements.
Jones WM, et al.
Journal of the American Chemical Society, 95(3), 826-835 (1973)
Dehydrogenation of amines. An approach to imines and aldehydes.
Trost BM and Liu GJ.
The Journal of Organic Chemistry, 46(22), 4617-4620 (1981)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service