Skip to Content
Merck
All Photos(1)

Key Documents

547905

Sigma-Aldrich

2,2′:5′,2′′:5′′,2′′′-Quaterthiophene

96%

Synonym(s):

α-Quarterthienyl, 4T

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€346.00

€346.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€346.00

About This Item

Empirical Formula (Hill Notation):
C16H10S4
CAS Number:
Molecular Weight:
330.51
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

€346.00


Check Cart for Availability

Request a Bulk Order

Assay

96%

mp

211-214 °C (lit.)

semiconductor properties

P-type (mobility=1.2×10−3 cm2/V·s)

SMILES string

c1csc(c1)-c2ccc(s2)-c3ccc(s3)-c4cccs4

InChI

1S/C16H10S4/c1-3-11(17-9-1)13-5-7-15(19-13)16-8-6-14(20-16)12-4-2-10-18-12/h1-10H

InChI key

FXEJOIFDICYSSO-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Katie M Lavigne et al.
Human brain mapping, 37(12), 4640-4653 (2016-08-02)
Schizophrenia patients show widespread impairments in brain activity during oddball tasks, which involve responding to infrequent target stimuli while refraining from responding during continuous non-target stimuli. In a network-based investigation comparing schizophrenia or schizoaffective patients to healthy controls, we sought
Taosheng Liu et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 38(2), 398-408 (2017-11-24)
Channel-encoding models offer the ability to bridge different scales of neuronal measurement by interpreting population responses, typically measured with BOLD imaging in humans, as linear sums of groups of neurons (channels) tuned for visual stimulus properties. Inverting these models to
Albert Jang et al.
Magnetic resonance in medicine, 79(2), 701-710 (2017-05-13)
To introduce a method of designing single and parallel transmit (pTx) 3D adiabatic π pulses for inverting and refocusing spins that are insensitive to transmit B A 3D adiabatic pulse is created by replacing each piece-wise constant element (or sub-pulse)
Jinjin Zhang et al.
Magnetic resonance in medicine, 75(2), 537-546 (2015-03-25)
Methods designed to image fast-relaxing spins, such as sweep imaging with Fourier transformation (SWIFT), often utilize high excitation bandwidth and duty cycle, and in some applications the optimal flip angle cannot be used without exceeding safe specific absorption rate (SAR)
Louis Van der Meeren et al.
iScience, 23(12), 101816-101816 (2020-12-11)
Regulated cell death (RCD) has a fundamental role in development, pathology, and tissue homeostasis. In order to understand the RCD mechanisms, it is essential to follow these processes in real time. Here, atomic force microscopy (AFM) is applied to morphologically

Articles

Oligothiophenes are important organic electronic materials which can be produced using synthetic intermediates and Suzuki coupling.

Thin, lightweight, and flexible electronic devices meet widespread demand for scalable, portable, and robust technology.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service