Skip to Content
Merck
All Photos(2)

Key Documents

547689

Sigma-Aldrich

Poly(4-vinylpyridine), cross-linked

Reillex® 402 ion-exchange resin, 2 % cross-linked with divinylbenzene

Synonym(s):

4-Vinylpyridine-divinylbenzene copolymer, Divinylbenzene-4-vinylpyridine copolymer

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
€312.00

€312.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 G
€312.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

€312.00


Check Cart for Availability

Request a Bulk Order

Quality Level

crosslinking

2 % cross-linked with divinylbenzene

manufacturer/tradename

Reillex® 402 ion-exchange resin

SMILES string

n2ccc(cc2)C=C.c1(c(cccc1)C=C)C=C

InChI

1S/C10H10.C7H7N/c1-3-9-7-5-6-8-10(9)4-2;1-2-7-3-5-8-6-4-7/h3-8H,1-2H2;2-6H,1H2

InChI key

HUEZUDXJCGHIHG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Reillex is a registered trademark of Vertellus Specialties, Inc.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service