Skip to Content
Merck
All Photos(1)

Key Documents

547522

Sigma-Aldrich

StratoSpheres PL-MPH resin

50-100 mesh, extent of labeling: 3.0-4.0 mmol/g loading, 1 % cross-linked

Synonym(s):

Morpholine, polymer-bound

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€169.00

€169.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€169.00

About This Item

Empirical Formula (Hill Notation):
C29H33NO
CAS Number:
Molecular Weight:
411.58
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.22

€169.00


Check Cart for Availability

Request a Bulk Order

product line

StratoSpheres

crosslinking

1 % cross-linked

reaction suitability

reaction type: solution phase peptide synthesis
reactivity: proton reactive

extent of labeling

3.0-4.0 mmol/g loading

particle size

50-100 mesh

functional group

amine

SMILES string

N1CCOCC1

InChI

1S/C4H9NO/c1-3-6-4-2-5-1/h5H,1-4H2

InChI key

YNAVUWVOSKDBBP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

StratoSpheres is a trademark of Agilent Technologies, Inc.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Benoît Pilmis et al.
The Journal of antimicrobial chemotherapy, 70(1), 14-22 (2014-09-11)
Antifungal prescription remains a challenge in pregnant women because of uncertainties regarding fetal toxicity and altered maternal pharmacokinetic parameters that may affect efficacy or increase maternal and fetal toxicity. We present updated data reviewing the available knowledge and current recommendations
Avinash Bhadani et al.
Physical chemistry chemical physics : PCCP, 17(29), 19474-19483 (2015-07-07)
A new series of ester functionalized cationic gemini surfactants having different cationic headgroups (i.e. piperidinium, pyrrolidinium, morpholinium and quaternary ammonium) have been synthesized and characterized using NMR and Mass spectroscopy. These new gemini surfactants were investigated for their micellization and
Xing Liu et al.
Biochemical pharmacology, 97(1), 111-121 (2015-07-28)
Gefitinib (GEF), an inhibitor of epidermal growth factor receptor (EGFR) tyrosine kinase, is widely used for the treatment of cancers, particularly non-small cell lung cancer. However, its clinical use is limited by multiple adverse effects associated with GEF, such as
Alfonso Pérez-Gallardo et al.
Journal of food science and technology, 52(9), 5601-5610 (2015-09-08)
There is increased interest in berry fruits due to health benefits, and maintenance of fruit quality for longer periods of time has been a priority. We previously found that starch based coatings applied on raspberries was associated to volatile compounds
Almeqdad Y Habashneh et al.
Archiv der Pharmazie, 347(6), 415-422 (2014-03-13)
A new series of N1-(flavon-6-yl)amidrazones were synthesized by reacting the hydrazonoyl chloride derived from 6-aminoflavone with the appropriate sec-cyclic amines. The antitumor activities of these compounds were evaluated on breast cancer (MCF-7) and leukemic (K562) cell lines. Among the compounds

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service