Skip to Content
Merck
All Photos(1)

Key Documents

546321

Sigma-Aldrich

2,6-Diphenylisonicotinic acid

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H13NO2
CAS Number:
Molecular Weight:
275.30
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

283-286 °C (lit.)

SMILES string

OC(=O)c1cc(nc(c1)-c2ccccc2)-c3ccccc3

InChI

1S/C18H13NO2/c20-18(21)15-11-16(13-7-3-1-4-8-13)19-17(12-15)14-9-5-2-6-10-14/h1-12H,(H,20,21)

InChI key

PWKUURSWFJBXGO-UHFFFAOYSA-N

General description

2,6-Diphenylisonicotinic acid can be synthesized by reacting ethyl 2,6-diphenylisonicotinate in ethanol with KOH solution.

Application

2,6-Diphenylisonicotinic acid may be used as one of the reactants in the synthesis of 7-substituted spiro[chroman-2,4′-piperidin]-4-one derivatives and ethyl 2,6-diphenylisonicotinate (a tridentate ligand).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of spiro [chroman-2, 4'-piperidin]-4-one derivatives as acetyl-CoA carboxylase inhibitors.
Shinde P, et al.
Bioorganic & Medicinal Chemistry Letters, 19(3), 949-953 (2009)
Studies with ?-oxoalkanonitriles: Simple novel synthesis of 3-[2, 6-diaryl-4-pyridyl]-3-oxopropanenitriles.
Riyadh SM, et al.
Molecules (Basel), 13(12), 3140-3148 (2008)
Synthesis of alkynylgold (III) complexes with bis-cyclometalating ligand derived from ethyl 2, 6-diphenylisonicotinate and their structural, electrochemical, photo-and electroluminescence studies.
Au VKM, et al.
Journal of Organometallic Chemistry, 792, 109-116 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service