Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

544213

Sigma-Aldrich

1,4-Diethoxybenzene

97%

Synonym(s):

Hydroquinone diethylether

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

Linear Formula:
(C2H5O)2C6H4
CAS Number:
Molecular Weight:
166.22
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

mp

69-72 °C (lit.)

SMILES string

CCOc1ccc(OCC)cc1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Karen A Nolan et al.
Bioorganic & medicinal chemistry letters, 16(24), 6246-6254 (2006-10-03)
From in silico docking and COMPARE analysis, novel inhibitors of human NAD(P)H quinone oxidoreductase (NQO1) have been identified from the NCI compound database, the most potent of which has an observed IC(50) of 0.7muM. The inhibitors exhibit a diverse range

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service