Skip to Content
Merck
All Photos(1)

Key Documents

543985

Sigma-Aldrich

Propargyl benzoate

98%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€61.10

€61.10


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€61.10

About This Item

Linear Formula:
C6H5CO2CH2C≡CH
CAS Number:
Molecular Weight:
160.17
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€61.10


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

98%

form

liquid

refractive index

n20/D 1.5320 (lit.)

bp

225-226 °C (lit.)

density

1.106 g/mL at 25 °C (lit.)

functional group

ester
phenyl

SMILES string

O=C(OCC#C)c1ccccc1

InChI

1S/C10H8O2/c1-2-8-12-10(11)9-6-4-3-5-7-9/h1,3-7H,8H2

InChI key

NBDHEMWCIUHARG-UHFFFAOYSA-N

General description

Propargyl benzoate is an aromatic ester containing a terminal acetylene group. It can be synthesized by reacting propargyl bromide and benzoyl chloride.[1]

Application

Propargyl benzoate may be used in the preparation of amphiphilic graft copolymers of poly(ε-caprolactone) (PCL).[2]

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Combination of ring-opening polymerization and ?click chemistry?: toward functionalization and grafting of poly (?-caprolactone).
Riva R, et al.
Macromolecules, 40(4), 796-803 (2007)
Nickel-Catalyzed Tandem Coupling of ?,?-Enones, Alkynes, and Alkynyltins for the Regio-and Stereoselective Synthesis of Conjugated Enynes.
Ikeda S, et al.
The Journal of Organic Chemistry, 61(23), 8248-8255 (1996)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service