Skip to Content
Merck
All Photos(1)

Key Documents

543144

Sigma-Aldrich

6-Chloro-1-hexene

96%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl(CH2)4CH=CH2
CAS Number:
Molecular Weight:
118.60
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Assay:
96%
Pricing and availability is not currently available.

Quality Level

Assay

96%

refractive index

n20/D 1.438 (lit.)

bp

135-136 °C (lit.)

density

0.896 g/mL at 25 °C (lit.)

functional group

alkyl halide
allyl
chloro

SMILES string

ClCCCCC=C

InChI

1S/C6H11Cl/c1-2-3-4-5-6-7/h2H,1,3-6H2

InChI key

BLMIXWDJHNJWDT-UHFFFAOYSA-N

General description

6-Chloro-1-hexene is an ω-chloro-1-alkene.[1] It reacts with β-substituted imines in the presence of a rhodium catalyst to afford tri- and tetra-substituted imines.[2] It undergoes cross-coupling reaction in the presence of copper(II) chloride and 1-phenylpropyne.[3]

Application

6-Chloro-1-hexene may be used in the synthesis of 1-(5-hexen)-2,3-dimethyl-imidazolium chloride.[4]

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

84.0 °F - closed cup

Flash Point(C)

28.9 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Stereoselective alkylation of a, ?-unsaturated imines via CH bond activation
Colby, Denise A., Robert G. Bergman, and Jonathan A. Ellman
Journal of the American Chemical Society, 128.17, 5604-5605 (2006)
The Signs of Si29-H1 and Si29-F19 Coupling Constants
Danyluk SS.
Journal of the American Chemical Society, 86.20, 4504-4505 (1964)
New ionic liquid-modified silica gels as recyclable materials for l-proline-or H?Pro?Pro?Asp?NH2-catalyzed aldol reaction
Aprile C, et al.
Green Chemistry, 9.12, 1328-1334 (2007)
Copper nanoparticle-catalyzed cross-coupling of alkyl halides with Grignard reagents.
Kim JH and Chung YK.
Chemical Communications (Cambridge, England), 49(94), 11101-11103 (2013)
Christopher L Rock et al.
Dalton transactions (Cambridge, England : 2003), 48(2), 461-467 (2018-11-30)
The phosphine-substituted α-diimine Ni precursor, (Ph2PPrDI)Ni, has been found to catalyze alkene hydrosilylation in the presence of Ph2SiH2 with turnover frequencies of up to 124 h-1 at 25 °C (990 h-1 at 60 °C). Moreover, the selective hydrosilylation of allylic

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service