Skip to Content
Merck
All Photos(1)

Key Documents

542806

Sigma-Aldrich

4-Pyridylamidoxime

97%

Synonym(s):

Isonicotinamide oxime

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H7N3O
CAS Number:
Molecular Weight:
137.14
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

128-133 °C (lit.)

functional group

amine
oxime

SMILES string

N\C(=N/O)c1ccncc1

InChI

1S/C6H7N3O/c7-6(9-10)5-1-3-8-4-2-5/h1-4,10H,(H2,7,9)

InChI key

ZUUATXHRJFHSGO-UHFFFAOYSA-N

Application

4-Pyridylamidoxime may be used in the synthesis of substituted α-1,2,4-oxadiazolo esters.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A 'one-pot'synthesis of a-1, 2, 4-oxadiazolo esters from malonic diesters and amidoximes under solvent-free conditions.
Du W, et al.
Tetrahedron Letters, 47(25), 4271-4274 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service