Skip to Content
Merck
All Photos(1)

Documents

542016

Sigma-Aldrich

5-Bromo-2-hydroxybenzophenone

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5C(O)C6H3(Br)OH
CAS Number:
Molecular Weight:
277.11
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

98%

mp

111-114 °C (lit.)

SMILES string

Oc1ccc(Br)cc1C(=O)c2ccccc2

InChI

1S/C13H9BrO2/c14-10-6-7-12(15)11(8-10)13(16)9-4-2-1-3-5-9/h1-8,15H

InChI key

IVIICRNXAGUXLR-UHFFFAOYSA-N

General description

5-Bromo-2-hydroxybenzophenone (HBBP) can be synthesized by employing p-bromophenyl benzoate as a starting material.

Application

5-Bromo-2-hydroxybenzophenone may be used:
  • As a raw material for the synthesis of various monocondensed Schiff bases.
  • In the preparation of 4-bromo-2-{a-[1-(2-thienyl) benzimidazol-2-yl] benzyl} phenol.
  • As a reactant in the synthesis of 4-bromo-2-{[2-(2-hydroxybenzylideneamino)phenylimino] phenylmethyl} phenol.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

(5-Bromo-2-hydroxyphenyl)(phenyl) methanone.
Yang FK, et al.
Acta Crystallographica Section B, Structural Science, Crystal Engineering and Materials, 64(10), o1873-o1873 (2008)
4-Bromo-2-{a-[1-(2-thienyl) benzimidazol-2-yl] benzyl} phenol.
Yang FK, et al.
Acta Crystallographica Section B, Structural Science, Crystal Engineering and Materials, 63(6), o3041-o3041 (2007)
4-Bromo-2-{[2-(2-hydroxybenzylideneamino) phenylimino] phenylmethyl} phenol.
Yang FK, et al.
Acta Crystallographica Section B, Structural Science, Crystal Engineering and Materials, 63(12), o4608-o4608 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service