Skip to Content
Merck
All Photos(1)

Key Documents

541249

Sigma-Aldrich

Copper(II) molybdate

99%

Synonym(s):

Copper molybdate, Cupric molybdate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CuMoO4
CAS Number:
Molecular Weight:
223.48
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
Assay:
99%
form:
powder

Assay

99%

form

powder

mp

820 °C (lit.)

application(s)

battery manufacturing

SMILES string

[Cu++].[O-][Mo]([O-])(=O)=O

InChI

1S/Cu.Mo.4O/q+2;;;;2*-1

InChI key

IKUPISAYGBGQDT-UHFFFAOYSA-N

General description

Copper molybdate CuMoO4 can exists in two polymorphic forms, stable-α form and metastable-γ form. It exhibits properties such as thermochromic, trisochromic and piezochromic. Pure α-phase copper molybdate can be prepared by the sol-gel method using citric acid, ammonium heptamolybdate (NH4)6Mo7O24 . 4H2O, copper salts CuX2nH2O (X=NO3- ), Cl- and CH3COO-.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sol?gel synthesis and sintering of submicronic copper molybdate (?-CuMoO4) powders
Benchikhi M, et al.
Ceramics International, 40(4), 5371-5377 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service