Skip to Content
Merck
All Photos(1)

Key Documents

540862

Sigma-Aldrich

2,4-Dichlorobenzenethiol

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl2C6H3SH
CAS Number:
Molecular Weight:
179.07
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.619 (lit.)

bp

60-62 °C/1.5 mmHg (lit.)

density

1.418 g/mL at 25 °C (lit.)

SMILES string

Sc1ccc(Cl)cc1Cl

InChI

1S/C6H4Cl2S/c7-4-1-2-6(9)5(8)3-4/h1-3,9H

InChI key

FGBVJFREPSJSNG-UHFFFAOYSA-N

General description

2,4-Dichlorobenzenethiol is a halogenated aromatic thiol.

Application

2,4-Dichlorobenzenethiol (24DBT) may be used in the following:
  • Preparation of hyperbranched poly(phenylene sulfide) (HPPS).
  • Functionalization of printed gold nanoparticle films.
  • To synthesize N,N-dialkyl(aminopropyl)amino diaryl sulphide.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G Liu et al.
Journal of medicinal chemistry, 43(21), 4025-4040 (2000-10-29)
The interaction between leukocyte function-associated antigen-1 (LFA-1), a member of the beta(2)-integrin family of adhesion molecules, and intracellular adhesion molecule ICAM-1 (cd54) is thought to play a critical role in the inflammatory process. On the basis of an anilino diaryl
Cunfa Sun et al.
ACS nano, 13(5), 5975-5986 (2019-05-09)
Copper-hydrides are known catalysts for several technologically important reactions such as hydrogenation of CO, hydroamination of alkenes and alkynes, and chemoselective hydrogenation of unsaturated ketones to unsaturated alcohols. Stabilizing copper-based particles by ligand chemistry to nanometer scale is an appealing
Hyperbranched Polyphenylene Sulfide as Stabilizer to Prepare Colloidal Silver Nanoparticles.
Kuang J, et al.
Asian Journal of Chemistry, 27(11), 4034-4034 (2015)
A Feasible Synthetic Route for Linear PTHF-Hyperbranched Poly (phenyl sulfide) Block Copolymers.
Zhu Z and Pan C.
Macromolecular Chemistry and Physics, 208(12), 1274-1282 (2007)
Esma Khatun et al.
Nanoscale, 9(24), 8240-8248 (2017-06-06)
We report the synthesis of a new silver cluster, [Ag

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service