Skip to Content
Merck
All Photos(2)

Key Documents

540129

Sigma-Aldrich

(2S)-(+)-Glycidyl tosylate

98%

Synonym(s):

(S)-(+)-Glycidyl p-toluenesulfonate, (S)-(+)-Oxirane-2-methanol p-toluenesulfonate

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€36.80

€36.80


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€36.80

About This Item

Empirical Formula (Hill Notation):
C10H12O4S
CAS Number:
Molecular Weight:
228.26
Beilstein:
3592143
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

€36.80


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

98%

mp

46-49 °C (lit.)

functional group

ether
tosylate

storage temp.

2-8°C

SMILES string

Cc1ccc(cc1)S(=O)(=O)OC[C@@H]2CO2

InChI

1S/C10H12O4S/c1-8-2-4-10(5-3-8)15(11,12)14-7-9-6-13-9/h2-5,9H,6-7H2,1H3/t9-/m0/s1

InChI key

NOQXXYIGRPAZJC-VIFPVBQESA-N

Looking for similar products? Visit Product Comparison Guide

Application

Used in the first enantioselective synthesis of (R)- and (S)-4-acetyl-3-(hydroxymethyl)-3,4-dihydro-2H-pyrido[3,2-b]oxazines.[1]

Legal Information

Manufactured under license by Sterling Pharma Solutions Limited, using Jacobsen HKR technology.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Carc. 1B - Eye Dam. 1 - Muta. 2 - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Henry, N. et al.
Tetrahedron Letters, 45, 1465-1465 (2004)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service