Skip to Content
Merck
All Photos(1)

Key Documents

539317

Sigma-Aldrich

n-Propyltriethoxysilane

97%

Synonym(s):

Triethoxypropylsilane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5O)3SiCH2CH2CH3
CAS Number:
Molecular Weight:
206.35
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

97%

form

liquid

refractive index

n20/D 1.396 (lit.)

bp

179-180 °C (lit.)

density

0.892 g/mL at 25 °C (lit.)

SMILES string

CCC[Si](OCC)(OCC)OCC

InChI

1S/C9H22O3Si/c1-5-9-13(10-6-2,11-7-3)12-8-4/h5-9H2,1-4H3

InChI key

NBXZNTLFQLUFES-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

n-Propyltriethoxysilane (PTES) is a mesoporous hyperbranched organosilane that can be used as a grafting agent. It can be used in the functionalization of a variety of substrates by attaching the silane groups with the surface atoms.[1][2]

Application

n-Propyltriethoxysilane was used for surface modification of titania particles[3] and nanotubes.[4] Propyltriethoxysilane may be used as an organosilicon precursor to prepare O2 sensor platform.[5] They were also used to coat mesoporous silica tablets by the sol-gel technique.[6]

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

137.3 °F - closed cup

Flash Point(C)

58.5 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chemical functionalisation of titania nanotubes and their utilisation for the fabrication of reinforced polystyrene composites.
Journal of Materials Chemistry, 17(22), 2351-2358 (2007)
Synthesis of mesoporous silica materials functionalized with n-propyl groups
Lesaint C, et al.
Microporous and Mesoporous Materials : The Official Journal of the International Zeolite Association, 83(1-3), 76-84 (2005)
Effects of surface coating on the controlled release of vitamin B1 from mesoporous silica tablets
Journal of Controlled Release : Official Journal of the Controlled Release Society, 119(2), 215-221 (2007)
Novel hybrid optical sensor materials for in-breath O 2 analysis
Analyst, 133(2), 241-247 (2008)
The effect of surface modification with silane coupling agent on suppressing the photo-catalytic activity of fine TiO2 particles as inorganic UV filter
Applied Surface Science, 254(2), 563-569 (2007)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service