Skip to Content
Merck
All Photos(1)

Key Documents

538833

Sigma-Aldrich

Poly(1-hexadecene-sulfone)

average Mw ~10,000,000, powder

Synonym(s):

BIOBLAND®-16

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12162002

form

powder

mol wt

average Mw ~10,000,000

impurities

≤0.2 wt. % Monomer

mp

50.4 °C (lit.)

solubility

H2O: insoluble

density

0.985 g/mL at 25 °C (lit.)

Application

Studied for use in medical devices, sensors requiring a gas permeable membrane, electron beam resist lithography.

Features and Benefits

High permeability to oxygen and nitrogen. Good biocompatibility.
Hydrophobic, biocompatible polymer.

Legal Information

BIOBLAND is a registered trademark of Affymetrix, Inc.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service