Skip to Content
Merck
All Photos(1)

Key Documents

53815

Sigma-Aldrich

4-Methoxybenzyl carbazate

≥98.0%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H12N2O3
CAS Number:
Molecular Weight:
196.20
Beilstein:
2052164
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98.0%

mp

75-78 °C

SMILES string

COc1ccc(COC(=O)NN)cc1

InChI

1S/C9H12N2O3/c1-13-8-4-2-7(3-5-8)6-14-9(12)11-10/h2-5H,6,10H2,1H3,(H,11,12)

InChI key

JKBMMHKEAGEILO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

4-Methoxybenzyl carbazate may be used to synthesize 5,10-diiminoporphodimethene (a dearomatized porphyrinoid) and N-(2,4-dihydroxythiobenzoyl)-4-methoxybenzyl carbazate

Other Notes

Starting material for the azide used to introduce the pMZ-amino protection which is removable under mild hydrolytic conditions

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and fungistatic activity of new groups of 2, 4-dihydroxythiobenzoyl derivatives against phytopathogenic fungi
Legocki, Jan, et al.
Journal of Agricultural and Food Chemistry, 51.2 , 362-368 (2003)
C. Ressler et al.
The Journal of Organic Chemistry, 36, 3961-3961 (1971)
Experimental and Theoretical Characterization of 5, 10-Diminoporphodimethenes: Dearomatized Porphyrinoids from Palladium-Catalyzed Hydrazinations of 5, 10-Diarylporphyrins.
Basic B, et al.
ChemPlusChem, 79(6), 813-824 (2014)
H. Yamada et al.
Bulletin of the Chemical Society of Japan, 57, 3333-3333 (1984)
F. Weygand et al.
Chemische Berichte, 95, 1-1 (1962)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service