Skip to Content
Merck

53675

α-Humulene

≥96.0% (GC)

Synonym(s):

alpha-Humulene, α-Caryophyllene, trans,trans,trans-2,6,6,9-Tetramethyl-1,4,8-cycloundecatriene

Sign In to View Organizational & Contract Pricing.

About This Item

Empirical Formula (Hill Notation):
C15H24
CAS Number:
Molecular Weight:
204.35
UNSPSC Code:
12352100
NACRES:
NA.22
PubChem Substance ID:
EC Number:
229-816-7
Beilstein/REAXYS Number:
3240075
MDL number:
Assay:
≥96.0% (GC)

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

InChI key

FAMPSKZZVDUYOS-HRGUGZIWSA-N

InChI

1S/C15H24/c1-13-7-5-8-14(2)10-12-15(3,4)11-6-9-13/h6-7,10-11H,5,8-9,12H2,1-4H3/b11-6+,13-7+,14-10+

SMILES string

C\C1=C/CC(C)(C)\C=C\C\C(C)=C\CC1

assay

≥96.0% (GC)

refractive index

n20/D 1.503 (lit.)

bp

166-168 °C (lit.)

density

0.889 g/mL at 20 °C (lit.)

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

α-Humulene is a class of sesquiterpenes found in Cordia verbenacea essential oil.[1]

Application

α-Humulene can be used in the formation of secondary organic aerosol by oxidation.[2]

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

10 - Combustible liquids

wgk

WGK 3

flash_point_f

194.0 °F - closed cup

flash_point_c

90 °C - closed cup

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rafael C Dutra et al.
The Journal of pharmacy and pharmacology, 61(2), 243-250 (2009-01-31)
The objective of this work was to investigate the antiulcerogenic and anti-inflammatory activities of the essential oil from Pterodon emarginatus seeds. The following tests were used: ulcers induced by ethanol, indometacin and HCl/ethanol, and pleurisy induced by carrageenan in Swiss
Ping-Chuan Jiang et al.
Zhong yao cai = Zhongyaocai = Journal of Chinese medicinal materials, 32(8), 1224-1227 (2009-12-08)
To study the volatile oil of Murraya exotica. The volatile oil of Murraya exotica was extracted by steam-stilling and was identified by GC-MS-DS. More than 90 compounds were separated, and 59 compounds were identified, accounting for 93.9% of the total
João Paulo B Sousa et al.
Journal of pharmaceutical and biomedical analysis, 54(4), 653-659 (2010-11-26)
Copaifera species (Leguminoseae) are popularly known as "copaiba" or "copaíva". The oleoresins obtained from the trunk of these species have been extensively used in folk medicine and are commercialized in Brazil as crude oil and in several pharmaceutical and cosmetic
Ulrich Neuenschwander et al.
The Journal of organic chemistry, 77(6), 2865-2869 (2012-02-16)
Humulene is a sesquiterpene with an important biochemical lead structure, consisting of an 11-membered ring, containing three nonconjugated C═C double bonds, two of them being triply substituted and one being doubly substituted. As observed by many groups, one of the
Akinola O Ogunbinu et al.
Natural product communications, 4(3), 421-424 (2009-05-06)
The chemical composition of the essential oils from the leaves and stem bark of Eclipta prostrata (L.) L. and the aerial parts of Vernonia amygdalina Delile (Asteraceae) have been analyzed by capillary gas chromatography (GC) and gas chromatography/mass spectrometry (GC/MS).

Protocols

GC Analysis of Sweet Orange Essential Oil on SLB®-5ms (10 m x 0.10 mm I.D., 0.10 μm), Fast GC Analysis

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service